siRNA Details
| Gene Symbol: Rusc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037709.2 | NP_001032798.1 | 12 | 4945 | ||
| NM_199057.3 | NP_951012.2 | 12 | 4906 | |||
| GenBank mRNA | AK053542.1 | - | 6 | 4250 | ||
| AK122263.1 | - | 12 | 4825 | |||
| AK132170.1 | - | 12 | 4835 | |||
| AK166260.1 | - | 12 | 4840 | |||
| BC024790.1 | - | 9 | 2218 | |||
| BC141186.1 | - | 11 | 4670 | 9055798 | ||

Rusc2
Genomic Map