siRNA Details
| Gene Symbol: Rusc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037709.2 | NP_001032798.1 | 2 | 1169 | ||
| NM_199057.3 | NP_951012.2 | 2 | 1130 | |||
| GenBank mRNA | AK122263.1 | - | 2 | 1052 | ||
| AK132170.1 | - | 2 | 1059 | |||
| AK166260.1 | - | 2 | 1073 | |||
| BC050780.1 | - | 2 | 1081 | |||
| BC141186.1 | - | 1 | 903 | 9055798 | ||

Rusc2
Genomic Map