siRNA Details
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025572.1 | NP_001020743.1 | 9 | 1659 | ||
GenBank mRNA | AK134530.1 | - | 4 | 838 | ||
CF553222.1 | - | 1 | 81 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025572.1 | NP_001020743.1 | 9 | 1659 | ||
GenBank mRNA | AK134530.1 | - | 4 | 838 | ||
CF553222.1 | - | 1 | 81 |