siRNA Details
| Gene Symbol: Pdzd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033222.3 | NP_001028394.2 | 5 | 3510 | ||
| GenBank mRNA | AK040491.1 | - | 1 | 97 | ||
| AK145020.1 | - | 5 | 3510 | |||
| BC141370.1 | - | 5 | 3415 | 9055983 | ||
| BC141371.1 | - | 5 | 3415 | 9055984 | ||

Pdzd8
Genomic Map