siRNA Details
Gene Symbol: Gm14446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101605.1 | NP_001095075.1 | 2 | 1822 | ||
NM_001110517.1 | NP_001103987.1 | 2 | 1903 | |||
GenBank mRNA | AK090072.1 | - | 3 | 1843 |
Gene Symbol: 2010002M12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053217.2 | NP_444447.1 | 3 | 1956 | ||
GenBank mRNA | AK136525.1 | - | 2 | 1808 | ||
AK153247.1 | - | 3 | 1957 | |||
BC003493.1 | - | 2 | 773 | |||
BC138202.1 | - | 2 | 1787 | 8861221 |
