siRNA Details
| Gene Symbol: Krtap5-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037822.1 | NP_001032911.1 | 1 | 965 | ||
| GenBank mRNA | BC111113.1 | - | 1 | 965 | 40051363 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap5-5| Gene Symbol: Krtap5-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037822.1 | NP_001032911.1 | 1 | 965 | ||
| GenBank mRNA | BC111113.1 | - | 1 | 965 | 40051363 | |