siRNA Details
| Gene Symbol: Krtap5-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037822.1 | NP_001032911.1 | 1 | 1126 | ||
| GenBank mRNA | AK020690.1 | - | 3 | 947 | ||
| BC111113.1 | - | 1 | 1126 | 40051363 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap5-5| Gene Symbol: Krtap5-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037822.1 | NP_001032911.1 | 1 | 1126 | ||
| GenBank mRNA | AK020690.1 | - | 3 | 947 | ||
| BC111113.1 | - | 1 | 1126 | 40051363 | ||