siRNA Details
| Gene Symbol: Gucy2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127318.1 | NP_001120790.1 | 27 | 3342 | ||
| NM_145067.3 | NP_659504.2 | 26 | 3270 | |||
| GenBank mRNA | AK136496.1 | - | 27 | 3366 | ||
| BC034064.1 | - | 20 | 2252 | |||
| BC099968.1 | - | 26 | 3270 | 6432585 | ||

Gucy2c
Genomic Map