siRNA Details
Gene Symbol: H60a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010400.2 | NP_034530.1 | 3013 | |||
GenBank mRNA | AK169909.1 | - | 3015 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: H60a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010400.2 | NP_034530.1 | 3013 | |||
GenBank mRNA | AK169909.1 | - | 3015 |