siRNA Details
| Gene Symbol: H60a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010400.2 | NP_034530.1 | 488 | |||
| GenBank mRNA | AF084643.1 | - | 297 | |||
| AK166223.1 | - | 445 | ||||
| AK169909.1 | - | 490 | ||||
| AK170137.1 | - | 461 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H60a| Gene Symbol: H60a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010400.2 | NP_034530.1 | 488 | |||
| GenBank mRNA | AF084643.1 | - | 297 | |||
| AK166223.1 | - | 445 | ||||
| AK169909.1 | - | 490 | ||||
| AK170137.1 | - | 461 | ||||