siRNA Details
| Gene Symbol: Inhbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008381.3 | NP_032407.1 | 2 | 3771 | ||
| GenBank mRNA | AK132352.1 | - | 2 | 3773 | ||
| BC024067.1 | - | 1 | 1923 | |||
| BC048845.1 | - | 2 | 2476 | |||
| X69620.1 | - | 1 | 1737 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inhbb| Gene Symbol: Inhbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008381.3 | NP_032407.1 | 2 | 3771 | ||
| GenBank mRNA | AK132352.1 | - | 2 | 3773 | ||
| BC024067.1 | - | 1 | 1923 | |||
| BC048845.1 | - | 2 | 2476 | |||
| X69620.1 | - | 1 | 1737 | |||