siRNA Details
| Gene Symbol: Cfp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008823.3 | NP_032849.2 | 7 | 1137 | ||
| GenBank mRNA | AK150212.1 | - | 7 | 1098 | ||
| AK152782.1 | - | 7 | 1088 | |||
| AK154306.1 | - | 7 | 1122 | |||
| AK159291.1 | - | 7 | 1138 | |||
| AK159609.1 | - | 7 | 1088 | |||
| AK159853.1 | - | 7 | 1088 | |||
| AK171374.1 | - | 7 | 1121 | |||
| BC132369.1 | - | 7 | 1121 | 40130646 | ||
| BC138306.1 | - | 7 | 1121 | 8861326 | ||
| X12905.1 | - | 6 | 986 | |||

Cfp
Genomic Map