siRNA Details
| Gene Symbol: Slit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011412.3 | NP_035542.2 | 2 | 288 | ||
| GenBank mRNA | AF144629.1 | - | 2 | 229 | ||
| AK147292.1 | - | 2 | 289 | |||
| BC150780.1 | - | 2 | 265 | 9087691 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slit3| Gene Symbol: Slit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011412.3 | NP_035542.2 | 2 | 288 | ||
| GenBank mRNA | AF144629.1 | - | 2 | 229 | ||
| AK147292.1 | - | 2 | 289 | |||
| BC150780.1 | - | 2 | 265 | 9087691 | ||