siRNA Details
| Gene Symbol: Fancd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033244.3 | NP_001028416.2 | 23 | 2243 | ||
| GenBank mRNA | AK142384.1 | - | 23 | 2241 | ||
| AK155602.1 | - | 23 | 2222 | |||
| BC040776.1 | - | 11 | 1124 | |||
| BC042619.1 | - | 11 | 1124 | |||
| BC139004.1 | - | 23 | 2241 | 8862026 | ||

Fancd2
Genomic Map