siRNA Details
| Gene Symbol: Klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033531.3 | NP_001028703.2 | 10 | 2273 | ||
| NM_001163020.1 | NP_001156492.1 | 11 | 2372 | |||
| GenBank mRNA | AK134944.1 | - | 10 | 2245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl32| Gene Symbol: Klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033531.3 | NP_001028703.2 | 10 | 2273 | ||
| NM_001163020.1 | NP_001156492.1 | 11 | 2372 | |||
| GenBank mRNA | AK134944.1 | - | 10 | 2245 | ||