siRNA Details
| Gene Symbol: Dos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195268.1 | NP_001182197.1 | 9 | 1844 | ||
| GenBank mRNA | AK035824.1 | - | 3 | 995 | ||
| AK137009.1 | - | 11 | 2181 | |||
| BC024795.1 | - | 1 | 1071 | |||
| BC026893.1 | - | 1 | 1285 | |||
| BC039779.1 | - | 1 | 1008 | |||
| BC052075.1 | - | 10 | 1898 | |||
| BC078442.1 | - | 5 | 1399 | |||
| BC100552.1 | - | 4 | 1227 | 5687195 | ||

Dos
Genomic Map