siRNA Details
| Gene Symbol: Cep170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024602.3 | NP_001019773.2 | 6 | 679 | ||
| GenBank mRNA | AK122236.1 | - | 7 | 608 | ||
| AK147410.1 | - | 6 | 620 | |||
| AK147612.1 | - | 4 | 284 | |||
| BC050077.1 | - | 6 | 513 | |||
| BC141174.1 | - | 5 | 479 | 9055786 | ||

Cep170b
Genomic Map