siRNA Details
| Gene Symbol: Cd27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033126.2 | NP_001028298.1 | 1, 2 | 275 | ||
| NM_001286753.1 | NP_001273682.1 | 1, 2 | 275 | |||
| GenBank mRNA | AA475779.1 | - | 1, 2 | 202 | ||
| AK137770.1 | - | 1, 2 | 252 | |||
| AK153895.1 | - | 1, 2 | 214 | |||
| AK154005.1 | - | 1, 2 | 259 | |||
| BC139125.1 | - | 1, 2 | 240 | 8862147 | ||
| BC139126.1 | - | 1, 2 | 240 | 8862148 | ||
| DT917063.1 | - | 1, 2 | 201 | |||
| DT924276.1 | - | 1, 2 | 201 | |||
| L24495.1 | - | 1, 2 | 317 | |||

Cd27
Genomic Map