siRNA Details
| Gene Symbol: Zfp217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033299.3 | NP_001028471.1 | 6 | 3708 | ||
| NM_001159683.1 | NP_001153155.1 | 7 | 3693 | |||
| GenBank mRNA | AK030249.1 | - | 5 | 2165 | ||
| AK139547.1 | - | 7 | 3695 | |||
| AK156475.1 | - | 6 | 3709 | |||
| AK163381.1 | - | 7 | 3700 | |||
| BC046393.1 | - | 3 | 702 | |||
| BC116329.1 | - | 5 | 3187 | 40096805 | ||
| BC116330.1 | - | 5 | 3187 | 40096806 | ||

Zfp217
Genomic Map