siRNA Details
| Gene Symbol: Zfp114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC096771.1 | - | 2 | 2876 | ||
| BC138311.1 | - | 2 | 2764 | 8861331 | ||
| BC138312.1 | - | 2 | 2764 | 8861332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp114| Gene Symbol: Zfp114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC096771.1 | - | 2 | 2876 | ||
| BC138311.1 | - | 2 | 2764 | 8861331 | ||
| BC138312.1 | - | 2 | 2764 | 8861332 | ||