siRNA Details
| Gene Symbol: Cog7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033318.3 | NP_001028490.2 | 2 | 371 | ||
| GenBank mRNA | AK030709.1 | - | 2 | 338 | ||
| AK145163.1 | - | 2 | 369 | |||
| AK154693.1 | - | 2 | 373 | |||
| AK163143.1 | - | 391 | ||||
| AK172200.1 | - | 2 | 373 | |||
| AK191299.1 | - | 2 | 230 | |||

Cog7
Genomic Map