siRNA Details
| Gene Symbol: Cog7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033318.3 | NP_001028490.2 | 8 | 1214 | ||
| GenBank mRNA | AK030709.1 | - | 8 | 1181 | ||
| AK145163.1 | - | 8 | 1212 | |||
| AK154693.1 | - | 8 | 1216 | |||
| AK163143.1 | - | 1234 | ||||
| AK172200.1 | - | 8 | 1216 | |||

Cog7
Genomic Map