siRNA Details
| Gene Symbol: Inpp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024617.3 | NP_001019788.1 | 25 | 3130 | ||
| NM_001297591.1 | NP_001284520.1 | 25 | 3107 | |||
| GenBank mRNA | AK139397.1 | - | 18 | 2607 | ||
| BC044876.1 | - | 11 | 1495 | |||
| BC064813.1 | - | 23 | 2832 | 6826283 | ||
| DQ296613.1 | - | 25 | 3107 | |||

Inpp4b
Genomic Map