siRNA Details
| Gene Symbol: OTUD7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020205.3 | NP_064590.2 | 12 | 2920 | ||
| XM_011509782.1 | XP_011508084.1 | 12 | 2944 | |||
| XM_011509783.2 | XP_011508085.1 | 12 | 3174 | |||
| XM_011509784.2 | XP_011508086.1 | 12 | 2738 | |||
| XM_011509785.2 | XP_011508087.1 | 12 | 6371 | |||
| XM_011509786.1 | XP_011508088.1 | 12 | 2810 | |||
| XM_011509787.1 | XP_011508089.1 | 12 | 2553 | |||
| XM_011509788.1 | XP_011508090.1 | 11 | 2662 | |||
| XM_017001850.1 | XP_016857339.1 | 12 | 2783 | |||
| XM_017001851.1 | XP_016857340.1 | 11 | 2769 | |||
| GenBank mRNA | AJ293573.1 | - | 12 | 2763 | ||
| AK299790.1 | - | 13 | 2819 | |||
| AL122102.1 | - | 2 | 1257 | |||
| BC072681.1 | - | 12 | 2857 | 6170513 | ||

OTUD7B
Genomic Map