siRNA Details
| Gene Symbol: A630095E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033325.2 | NP_001028497.1 | 3 | 265 | ||
| GenBank mRNA | AK042477.1 | - | 781 | |||
| AK144443.1 | - | 3 | 266 | |||
| BB804458.1 | - | 1 | 23 | |||
| BB873517.1 | - | 3 | 266 | |||
| CJ069767.1 | - | 3 | 266 | |||

A630095E13Rik
Genomic Map