siRNA Details
| Gene Symbol: Dhrsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033326.2 | NP_001028498.2 | 7 | 1667 | ||
| GenBank mRNA | AK030649.1 | - | 6 | 1595 | ||
| AK157461.1 | - | 7 | 1353 | |||
| BC138597.1 | - | 7 | 1310 | 8861617 | ||
| BC138599.1 | - | 7 | 1310 | 8861619 | ||

Dhrsx
Genomic Map