siRNA Details
| Gene Symbol: Tmtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033332.2 | NP_001028504.1 | 13 | 2520 | ||
| GenBank mRNA | AK158192.1 | - | 13 | 2508 | ||
| AK165838.1 | - | 13 | 2521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmtc3| Gene Symbol: Tmtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033332.2 | NP_001028504.1 | 13 | 2520 | ||
| GenBank mRNA | AK158192.1 | - | 13 | 2508 | ||
| AK165838.1 | - | 13 | 2521 | |||