siRNA Details
| Gene Symbol: Tmtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033332.2 | NP_001028504.1 | 5, 6 | 845 | ||
| NM_001110013.1 | NP_001103483.1 | 5, 6 | 845 | |||
| GenBank mRNA | AK044857.1 | - | 5, 6 | 801 | ||
| AK149759.1 | - | 5, 6 | 804 | |||
| AK158192.1 | - | 5, 6 | 831 | |||
| AK165838.1 | - | 5, 6 | 846 | |||

Tmtc3
Genomic Map