siRNA Details
| Gene Symbol: Bank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033350.2 | NP_001028522.2 | 6 | 1060 | ||
| GenBank mRNA | AK041071.1 | - | 3 | 318 | ||
| AK041242.1 | - | 6 | 1021 | |||
| AK156201.1 | - | 6 | 1070 | |||
| AY178734.1 | - | 6 | 1080 | |||
| BC026932.1 | - | 6 | 1007 | 4008261 | ||
| EU051378.1 | - | 5 | 661 | |||

Bank1
Genomic Map