siRNA Details
| Gene Symbol: Rasl10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013386.2 | NP_001013404.1 | 4 | 1515 | ||
| GenBank mRNA | AK045997.1 | - | 4 | 1285 | ||
| AK158941.1 | - | 4 | 1516 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasl10b| Gene Symbol: Rasl10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013386.2 | NP_001013404.1 | 4 | 1515 | ||
| GenBank mRNA | AK045997.1 | - | 4 | 1285 | ||
| AK158941.1 | - | 4 | 1516 | |||