siRNA Details
| Gene Symbol: Zfp865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033383.2 | NP_001028555.1 | 2 | 658 | ||
| NM_001290426.1 | NP_001277355.1 | 2 | 658 | |||
| GenBank mRNA | AK032361.1 | - | 1 | 45 | ||
| AK154737.1 | - | 2 | 659 | |||
| BC020373.1 | - | 2 | 449 | |||

Zfp865
Genomic Map