siRNA Details
| Gene Symbol: Cass4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080820.2 | NP_001074289.1 | 2 | 250 | ||
| NM_001276422.1 | NP_001263351.1 | 2 | 250 | |||
| NM_001276423.1 | NP_001263352.1 | 2 | 250 | |||
| GenBank mRNA | AK036419.1 | - | 2 | 224 | ||
| AK080960.1 | - | 2 | 224 | |||
| AK089450.1 | - | 2 | 250 | |||
| AK170726.1 | - | 2 | 250 | |||
| BC117858.1 | - | 2 | 144 | 40095182 | ||
| BC117859.1 | - | 2 | 143 | 40095183 | ||
| BC129975.1 | - | 2 | 141 | 40095184 | ||
| BC129976.1 | - | 2 | 143 | 40095185 | ||

Cass4
Genomic Map