siRNA Details
| Gene Symbol: Vprbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015507.2 | NP_001015507.1 | 5 | 485 | ||
| GenBank mRNA | AK122372.1 | - | 5 | 448 | ||
| BC065119.1 | - | 5 | 485 | 6853519 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vprbp| Gene Symbol: Vprbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015507.2 | NP_001015507.1 | 5 | 485 | ||
| GenBank mRNA | AK122372.1 | - | 5 | 448 | ||
| BC065119.1 | - | 5 | 485 | 6853519 | ||