siRNA Details
| Gene Symbol: Kcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029985.4 | NP_001025156.3 | 27 | 2995 | ||
| GenBank mRNA | AK154369.1 | - | 27 | 2983 | ||
| AY884211.1 | - | 27 | 2981 | |||
| BC059023.1 | - | 7 | 780 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcp| Gene Symbol: Kcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029985.4 | NP_001025156.3 | 27 | 2995 | ||
| GenBank mRNA | AK154369.1 | - | 27 | 2983 | ||
| AY884211.1 | - | 27 | 2981 | |||
| BC059023.1 | - | 7 | 780 | |||