siRNA Details
| Gene Symbol: Atxn7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033436.3 | NP_001028608.3 | 8 | 2105 | ||
| GenBank mRNA | AK036472.1 | - | 8 | 2054 | ||
| AK156016.1 | - | 8 | 2069 | |||
| AK158590.1 | - | 5 | 2250 | |||
| BC147479.1 | - | 8 | 2020 | 9056221 | ||
| BC147483.1 | - | 8 | 2020 | 9056229 | ||

Atxn7l1
Genomic Map