siRNA Details
| Gene Symbol: Abca17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031621.2 | NP_001026792.2 | 19 | 3156 | ||
| GenBank mRNA | AB112584.1 | - | 19 | 3156 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca17| Gene Symbol: Abca17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031621.2 | NP_001026792.2 | 19 | 3156 | ||
| GenBank mRNA | AB112584.1 | - | 19 | 3156 | ||