siRNA Details
| Gene Symbol: Tmem237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033449.1 | NP_001028621.1 | 11 | 1116 | ||
| NM_001037812.2 | NP_001032901.1 | 11 | 1075 | |||
| GenBank mRNA | AK133112.1 | - | 12 | 1100 | ||
| AK161697.1 | - | 11 | 1120 | |||
| AK167733.1 | - | 11 | 1077 | |||
| AK170118.1 | - | 11 | 1057 | |||
| BC147243.1 | - | 11 | 1095 | 9056304 | ||
| BC147244.1 | - | 11 | 1095 | 9056305 | ||
| CA465507.1 | - | 373 | ||||

Tmem237
Genomic Map