siRNA Details
| Gene Symbol: Tmem237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033449.1 | NP_001028621.1 | 9 | 868 | ||
| NM_001037812.2 | NP_001032901.1 | 9 | 827 | |||
| GenBank mRNA | AK133112.1 | - | 10 | 852 | ||
| AK161697.1 | - | 9 | 871 | |||
| AK167733.1 | - | 9 | 829 | |||
| AK170118.1 | - | 9 | 808 | |||
| BC147243.1 | - | 9 | 847 | 9056304 | ||
| BC147244.1 | - | 9 | 847 | 9056305 | ||
| BF722386.1 | - | 5 | 514 | |||
| CA465507.1 | - | 125 | ||||

Tmem237
Genomic Map