siRNA Details
| Gene Symbol: Slc22a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037749.2 | NP_001032838.1 | 9 | 1350 | ||
| GenBank mRNA | AK133291.1 | - | 9 | 1363 | ||
| BC100471.1 | - | 9 | 1350 | 30917628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a14| Gene Symbol: Slc22a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037749.2 | NP_001032838.1 | 9 | 1350 | ||
| GenBank mRNA | AK133291.1 | - | 9 | 1363 | ||
| BC100471.1 | - | 9 | 1350 | 30917628 | ||