siRNA Details
| Gene Symbol: A830080D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033472.2 | NP_001028644.2 | 11 | 2234 | ||
| GenBank mRNA | AK145159.1 | - | 10 | 2069 | ||
| BC075674.1 | - | 11 | 2196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A830080D01Rik| Gene Symbol: A830080D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033472.2 | NP_001028644.2 | 11 | 2234 | ||
| GenBank mRNA | AK145159.1 | - | 10 | 2069 | ||
| BC075674.1 | - | 11 | 2196 | |||