siRNA Details
| Gene Symbol: Bach2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144923.1 | - | 4 | 2457 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bach2os| Gene Symbol: Bach2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144923.1 | - | 4 | 2457 | ||