siRNA Details
| Gene Symbol: Specc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029936.3 | NP_001025107.1 | 2 | 1741 | ||
| NM_001281818.1 | NP_001268747.1 | 4 | 1958 | |||
| GenBank mRNA | AK050982.1 | - | 2 | 1742 | ||
| AK143663.1 | - | 4 | 1958 | |||
| AK220465.1 | - | 2 | 1668 | |||

Specc1
Genomic Map