siRNA Details
| Gene Symbol: Gm5535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033778.1 | NP_001028950.1 | 3 | 375 | ||
| GenBank mRNA | AK144672.1 | - | 3 | 375 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5535| Gene Symbol: Gm5535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033778.1 | NP_001028950.1 | 3 | 375 | ||
| GenBank mRNA | AK144672.1 | - | 3 | 375 | ||