siRNA Details
| Gene Symbol: Fbxo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015798.3 | NP_056613.1 | 3 | 326 | ||
| GenBank mRNA | AF176530.1 | - | 3 | 310 | ||
| AK082809.1 | - | 3 | 347 | |||
| AK163224.1 | - | 3 | 327 | |||
| AK166591.1 | - | 3 | 327 | |||
| AK166599.1 | - | 3 | 328 | |||
| AK166918.1 | - | 3 | 323 | |||
| AK167118.1 | - | 3 | 327 | |||
| AK167270.1 | - | 3 | 326 | |||

Fbxo15
Genomic Map