siRNA Details
| Gene Symbol: Fbxo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015798.3 | NP_056613.1 | 9 | 1260 | ||
| GenBank mRNA | AF176530.1 | - | 9 | 1244 | ||
| AK082809.1 | - | 9 | 1281 | |||
| AK163224.1 | - | 9 | 1261 | |||
| AK166591.1 | - | 9 | 1261 | |||
| AK166599.1 | - | 9 | 1262 | |||
| AK166918.1 | - | 9 | 2126 | |||
| AK167118.1 | - | 9 | 1261 | |||
| AK167270.1 | - | 9 | 1260 | |||

Fbxo15
Genomic Map