siRNA Details
| Gene Symbol: Fbxo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015798.3 | NP_056613.1 | 10 | 1571 | ||
| GenBank mRNA | AF176530.1 | - | 10 | 1555 | ||
| AK082809.1 | - | 10 | 1592 | |||
| AK163224.1 | - | 10 | 1572 | |||
| AK166591.1 | - | 10 | 1573 | |||
| AK166599.1 | - | 10 | 1574 | |||
| AK166918.1 | - | 10 | 2437 | |||
| AK167118.1 | - | 10 | 1573 | |||
| AK167270.1 | - | 10 | 1571 | |||

Fbxo15
Genomic Map