siRNA Details
Gene Symbol: Gm5796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029930.2 | NP_001025101.2 | 1582 | |||
GenBank mRNA | BC093494.1 | - | 1582 | 2812218 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029930.2 | NP_001025101.2 | 1582 | |||
GenBank mRNA | BC093494.1 | - | 1582 | 2812218 |