siRNA Details
| Gene Symbol: Mterf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158221.1 | - | 2 | 2337 | ||
| BC082778.1 | - | 2 | 1280 | 3994365 | ||
| BC094442.1 | - | 2 | 1289 | 3587753 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mterf1a| Gene Symbol: Mterf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158221.1 | - | 2 | 2337 | ||
| BC082778.1 | - | 2 | 1280 | 3994365 | ||
| BC094442.1 | - | 2 | 1289 | 3587753 | ||