siRNA Details
Gene Symbol: Psg27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037168.1 | NP_001032245.1 | 4 | 1157 | ||
GenBank mRNA | BC100351.1 | - | 4 | 1157 | 30713102 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psg27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037168.1 | NP_001032245.1 | 4 | 1157 | ||
GenBank mRNA | BC100351.1 | - | 4 | 1157 | 30713102 |