siRNA Details
| Gene Symbol: Rhox7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025086.2 | NP_001020257.2 | 5 | 519 | ||
| GenBank mRNA | DQ058644.1 | - | 519 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhox7a| Gene Symbol: Rhox7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025086.2 | NP_001020257.2 | 5 | 519 | ||
| GenBank mRNA | DQ058644.1 | - | 519 | |||